missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse EGF / Epidermal Growth Factor Protein
Recombinant Protein
Marca: enQuireBio™ QP4008-5mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_034243.2 | |
EGF / Epidermal Growth Factor Protein | |
Research Use Only | |
EGF | |
Recombinant Protein | |
E. coli | |
NSYPGCPSSY DGYCLNGGVC MHIESLDSYT CNCVIGYSGD RCQTRDLRWW ELR | |
Greater than 95% as determined by SDS-PAGE. |
13645 | |
5 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 250 pg/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 4 x 10^6 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |