missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse EPHA4 Protein
A cDNA sequence encoding the EPHA4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11797-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
EPHA4 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
VTGSRVYPAN EVTLLDSRSV QGELGWIASP LEGGWEEVSI MDEKNTPIRT YQVCNVMEAS QNNWLRTDWI TREGAQRVYI EIKFTLRDCN SLPGVMGTCK ETFNLYYYES DNDKERFIRE SQFGKIDTIA ADESFTQVDI GDRIMKLNTE IRDVGPLSKK GFYLAFQDVG ACIALVSVRV FYKKCPLTVR NLAQFPDTIT GADTSSLVEV RGSCVNNSEE KDVPKMYCGA DGEWLVPIGN CLCNAGHEEQ NGECQACKIG YYKALSTDAS CAKCPPHSYS VWEGATSCTC DRGFFRADND AASMPCTRPP SAPLNLISNV NETSVNLEWS SPQNTGGRQD ISYNVVCKKC GAGDPSKCRP CGSGVHYTPQ QNGLKTTRVS ITDLLAHTNY TFEIWAVNGV SKYNPSPDQS VSVTVTTNQA APSSIALVQA KEVTRYSVAL AWLEPDRPNG VILEYEVKYY EKDQNERSYR IVRTAARNTD IKGLNPLTSY VFHVRARTAA GYGDFSEPLE VTTNTVPSRI IGDGANSTLE HHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
The EPHA4 solution (0.5 mg/1ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |