missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Fas apoptotic inhibitory molecule 3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Fas apoptotic inhibitory molecule 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6017-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_081252.1 | |
Fas apoptotic inhibitory molecule 3 Protein | |
Research Use Only | |
Faim3 | |
Recombinant Protein | |
E. coli | |
RVLPEVQLNV EWGGSIIIEC PLPQLHVRMY LCRQMAKPGI CSTVVSNTFV KKEYERRVTL TPCLDKKLFL VEMTQLTEND DGIYACGVGM KTDKGKTQKI TLNVHNEYPE PFWEDEWTSE RPRWLHRFLQ HQMPWLHGSE HPSSSGVIAK VTTPAPKTEA PPVHQPSSIT SVTQHPRVYR AFSVSATKSP ALLPATTASK TSTQQAIRPL EASYSHHTRL HEQRTRHHGP HYGREDRGLH IPIPE | |
Greater than 90% as determined by SDS-PAGE. |
69169 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |