missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Glutathione S-transferase P 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Glutathione S-transferase P 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6126-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_038569.1 | |
Glutathione S-transferase P 1 Protein | |
Research Use Only | |
Gstp1 | |
Recombinant Protein | |
Yeast | |
PPYTIVYFPV RGRCEAMRML LADQGQSWKE EVVTIDTWMQ GLLKPTCLYG QLPKFEDGDL TLYQSNAILR HLGRSLGLYG KNQREAAQMD MVNDGVEDLR GKYVTLIYTN YENGKNDYVK ALPGHLKPFE TLLSQNQGGK AFIVGDQISF ADYNLLDLLL IHQVLAPGCL DNFPLLSAYV ARLSARPKIK AFLSSPEHVN RPINGNGKQ | |
Greater than 90% as determined by SDS-PAGE. |
14870 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |