missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ICAM4 / CD242 Protein
A DNA sequence encoding the Mus musculus (Mouse) ICAM4 / CD242, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8170-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_076381.1 | |
ICAM4 / CD242 Protein | |
Research Use Only | |
Icam4 | |
Recombinant Protein | |
E. coli | |
QQEWMQSPPA PSVTSAPFWV RLNPELEAVP PGGSAWLNCS HNCPLPVHSS LRTQLRQGKI VNGSGWVSYQ LLDVRAWNSK VRCVVTCAGE TREATARITA YKRPRSVILE PPVLVGHKYT LRCYVTHVFP VGFLVVSLRR GGRVIYHESL ERFTGSDLAN VTLTYVMRAG LNDLWQPLTC HARLNLDGLV VRSSSAPVML TVLALSPAS | |
Greater than 90% as determined by SDS-PAGE. |
78369 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |