missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IFNG / Interferon Gamma Protein
A DNA sequence encoding the Mus musculus (Mouse) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8841-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_032363.1 | |
IFNG / Interferon Gamma Protein | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
E. coli | |
IESLESLNNY FNSSGIDVEE KSLFLDIWRN WQKDGDMKIL QSQIISFYLR LFEVLKDNQA ISNNISVIES HLITTFFSNS KAKKDAFMSI AKFEVNNPQV QRQAFNELIR VVHQLLPESS LRKRKRSRC | |
Greater than 90% as determined by SDS-PAGE. |
15978 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |