missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL18BP Protein
A DNA sequence encoding the Mus musculus (Mouse) IL18BP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8880-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_034661.1 | |
IL18BP Protein | |
Research Use Only | |
IL18BP | |
Recombinant Protein | |
E. coli | |
TSAPQTTATV LTGSSKDPCS SWSPAVPTKQ YPALDVIWPE KEVPLNGTLT LSCTACSRFP YFSILYWLGN GSFIEHLPGR LKEGHTSREH RNTSTWLHRA LVLEELSPTL RSTNFSCLFV DPGQVAQYHI ILAQLWDGLK TAPSPSQETL SSHSPVSRSA GPGVA | |
Greater than 90% as determined by SDS-PAGE. |
16068 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |