missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Lymphocyte antigen 6E Protein
A DNA sequence encoding the Mus musculus (Mouse) Lymphocyte antigen 6E, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9800-ye-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001157508.1 | |
Lymphocyte antigen 6E Protein | |
Research Use Only | |
LY6E | |
Recombinant Protein | |
Yeast | |
LMCFSCTDQK NNINCLWPVS CQEKDHYCIT LSAAAGFGNV NLGYTLNKGC SPICPSENVN LNLGVASVNS YCCQSSFCNF SA | |
Greater than 90% as determined by SDS-PAGE. |
17069 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |