missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse M-CSF / CSF-1 Protein
Recombinant Protein
Marca: enQuireBio™ QP5408-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001107001.1 | |
M-CSF / CSF-1 Protein | |
Research Use Only | |
CSF1 | |
Recombinant Protein | |
E. coli | |
MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP | |
Greater than 95% as determined by SDS-PAGE. |
12977 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 10 ng/mL in the NFS-60 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |