missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Mannan-binding lectin serine protease 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Mannan-binding lectin serine protease 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10059-ba-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001003893.1 | |
Mannan-binding lectin serine protease 2 Protein | |
Research Use Only | |
Masp2 | |
Recombinant Protein | |
Baculovirus | |
SKWPEPVFGR LVSPGFPEKY ADHQDRSWTL TAPPGYRLRL YFTHFDLELS YRCEYDFVKL SSGTKVLATL CGQESTDTEQ APGNDTFYSL GPSLKVTFHS DYSNEKPFTG FEAFYAAEDV DECRVSLGDS VPCDHYCHNY LGGYYCSCRA GYVLHQNKHT CSALCSGQVF TGRSGYLSSP EYPQPYPKLS SCTYSIRLED GFSVILDFVE SFDVETHPEA QCPYDSLKIQ TDKGEHGPFC GKTLPPRIET DSHKVTITFA TDESGNHTGW KIHYTSTARP CPDPTAPPNG SISPVQAIYV LKDRFSVFCK TGFELLQGSV PLKSFTAVCQ KDGSWDRPMP ECSIIDCGPP DDLPNGHVDY ITGPEVTTYK AVIQYSCEET FYTMSSNGKY VCEADGFWTS SKGEKLPPVC EPVCGLSTHT IGGRIVGGQP AKPGDFPWQV LLLGQTTAAA GALIHDNWVL TAAHAVYEKR MAASSLNIRM GILKRLSPHY TQAWPEEIFI HEGYTHGAGF DNDIALIKLK NKVTINGSIM PVCLPRKEAA SLMRTDFTGT VAGWGLTQKG LLARNLMFVD IPIADHQKCT AVYEKLYPGV RVSANMLCAG LETGGKDSCR GDSGGALVFL DNETQRWFVG GIVSWGSINC GAADQYGVYT KVINYIPWIE NIISNF | |
Greater than 90% as determined by SDS-PAGE. |
17175 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |