missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MMP-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) MMP-3 Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8900-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_034939.1 | |
MMP-3 Protein | |
Research Use Only | |
MMP3 | |
Recombinant Protein | |
E. coli | |
PGSPKWRKSH ITYRIVNYTP DLPRQSVDSA IEKALKVWEE VTPLTFSRIS EGEADIMISF AVGEHGDFVP FDGPGTVLAH AYAPGPGING DAHFDDDERW TEDVTGTNLF LVAAHELGHS LGLYHSAKAE ALMYPVYKSS TDLSRFHLSQ DDVDGIQSLY GTPTASPDVL VVPTKSNSLE PETSPMCSST LFFDAVSTLR GEVLFFKDRH FWRKSLRTPE PEFYLISSFW PSLPSNMDAA YEVTNRDTVF IFKGNQFWAI RGHEELAGYP KSIHTLGLPA TVKKIDAAIS NKEKRKTYFF VEDKYWRFDE KKQSMEPGFP RKIAEDFPGV DSRVDAVFEA FGFLYFFSGS SQLEFDPNAK KVTHILKSNS WFNC | |
Greater than 90% as determined by SDS-PAGE. |
17392 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |