missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Oxysterol-binding protein-related protein 11 Protein
A DNA sequence encoding the Mus musculus (Mouse) Oxysterol-binding protein-related protein 11, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7816-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Oxysterol-binding protein-related protein 11 Protein | |
Research Use Only | |
Osbpl11 | |
Recombinant Protein | |
E. coli | |
MQGGEPASVM KVSESEGKLE GLATAVTPNK NSGNSSCGGA ISSSSSNSSR GGSAKGWQYS DHMESVNGYL MKYTNLVTGW QYRFFVLNNE AGLLEYFVNE QSRNQKPRGT LQLAGAVISP SDEDSHTFTV NAASGEQYKL RATDAKERQH WVSRLQICTQ HHTEAIGKNN PPLKSRSFSL ASSGNSPISQ RRPSQNAMSF FNVGHSKLQS VNKRAHLHPD HLVEVREMMS HAEGQQRDLI RRIECLPASG LLSSLDQDLL MLKATSMATM NCLNDCFHIL QLQHASHQKG ALPSGTTIEW LEPKIPLSNH YKNGAEQPFA TEPNKPMGAP EAQCVAESGV LAREPEDISA DDEVEDTCDN KEDDLGAVEE QRSVILHLLS QLKLGMDLTR VVLPTFILEK RSLLEMYADF MSHPDLFIGI TNGATPEDRM IRFVEYYLTS FHEGRKGAIA KKPYNPIIGE TFHCSWRMPK SEVASGVSSS SSTPAITDHA PLPEEAPTQS VSDCYTVRFV AEQVSHHPPV SGFYAECAER KMCVNAHVWT KSKFLGMSIG VTMVGEGVLC LLEHGEEYTF SLPCAYARSI LTVPWVELGG KVSVNCAKTG YSASITFHTK PFYGGKLHRV TAEVKYNLTN TVVCRVQGEW NSVLEFTYSN GETKFVDLAK LAVTKKRVRP LEKQDPFESR RLWKNVTDSL RESEIDKATE HKRSLEERQR TEERLRTETG TPWKTKYFIK EGDGWVYHKP LWKGIPSQPA E | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |