missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PGF1 Protein
A cDNA sequence encoding the PGF1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13028-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PGF1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
AGNNSTEVEV VPFNEVWGRS YCRPMEKLVY ILDEYPDEVS HIFSPSCVLL SRCSGCCGDE GLHCVPIKTA NITMQILKIP PNRDPHFYVE MTFSQDVLCE CRPILETTKA ERRKTKGKRK RSRNSQTEEP HPHHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
PGF1 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 0.1mM PMSF and 10% glycerol. |