missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein CYR61 Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein CYR61, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9095-ye-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_034646.1 | |
Protein CYR61 Protein | |
Research Use Only | |
CYR61 | |
Recombinant Protein | |
Yeast | |
TCPAACHCPL EAPKCAPGVG LVRDGCGCCK VCAKQLNEDC SKTQPCDHTK GLECNFGASS TALKGICRAQ SEGRPCEYNS RIYQNGESFQ PNCKHQCTCI DGAVGCIPLC PQELSLPNLG CPNPRLVKVS GQCCEEWVCD EDSIKDSLDD QDDLLGLDAS EVELTRNNEL IAIGKGSSLK RLPVFGTEPR VLFNPLHAHG QKCIVQTTSW SQCSKSCGTG ISTRVTNDNP ECRLVKETRI CEVRPCGQPV YSSLKKGKKC SKTKKSPEPV RFTYAGCSSV KKYRPKYCGS CVDGRCCTPL QTRTVKMRFR CEDGEMFSKN VMMIQSCKCN YNCPHPNEAS FRLYSLFNDI HKFRD | |
Greater than 90% as determined by SDS-PAGE. |
16007 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |