missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Retnlb Protein
Recombinant Protein
Marca: enQuireBio™ QP5475-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_076370.3 | |
Retnlb Protein | |
Research Use Only | |
Retnlb | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
57263 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA | |
Greater than 95% as determined by SDS-PAGE. |