missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100b Protein
A cDNA sequence encoding the S100b was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13405-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
S100b Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSELEKAMVA LIDVFHQYSG REGDKHKLKK SELKELINNE LSHFLEEIKE QEVVDKVMET LDEDGDGECD FQEFMAFVAM VTTACHEFFE HE | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
S100B Mouse solution containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |