missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VPS29 Protein
A cDNA sequence encoding the VPS29 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13944-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
VPS29 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMMLVLV LGDLHIPHRC NSLPAKFKKL LVPGKIQHIL CTGNLCTKES YDYLKTLAGD VHIVRGDFDE NLNYPEQKVV TVGQFKIGLI HGHQVIPWGD MASLALLQRQ FDVDILISGH THKFEAFEHE NKFYINPGSA TGAYNALETN IIPSFVLMDI QASTVVTYVY QLIGDDVKVE RIEYKKS | |
Greater than 95% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
The VPS29 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl, 1mM DTT and 10% glycerol. |