missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse WNT1-inducible-signaling pathway protein 1
A DNA sequence encoding the Mus musculus (Mouse) WNT1-inducible-signaling pathway protein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8274-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_058569.2 | |
WNT1-inducible-signaling pathway protein 1 | |
Research Use Only | |
WISP2 | |
Recombinant Protein | |
E. coli | |
QLCPAPCACP WTPPQCPPGV PLVLDGCGCC RVCARRLGES CDHLHVCDPS QGLVCQPGAG PSGRGAVCLF EEDDGSCEVN GRRYLDGETF KPNCRVLCRC DDGGFTCLPL CSEDVRLPSW DCPRPRRIQV PGRCCPEWVC DQAVMQPAIQ PSSAQGHQLS ALVTPASADG PCPNWSTAWG PCSTTCGLGI ATRVSNQNRF CQLEIQRRLC LSRPCLASRS HGSWNSAF | |
Greater than 90% as determined by SDS-PAGE. |
22403 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |