missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other AHSG Protein
A cDNA sequence encoding the AHSG was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10982-10ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
AHSG Protein | |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
MKSLVLLLCL AQLWGCHSAP HGPGLIYRQP NCDDPETEEA ALVAIDYINQ NLPWGYKHTL NQIDEVKVWP QQPSGELFEI EIDTLETTCH VLDPTPVARC SVRQLKEHAV EGDCDFQLLK LDGKFSVVYA KCDSSPDSAE DVRKVCQDCP LLAPLNDTRV VHAAKAALAA FNAQNNGSNF QLEEISRAQL VPLPPSTYVE FTVSGTDCVA KEATEAAKCN LLAEKQYGFC KATLSEKLGG AEVAVTCTVF QTQPVTSQPQ PEGANEAVPT PVVDPDAPPS PPLGAPGLPP AGSPPDSHVL LAAPPGHQLH RAHYDLRHTF MGVVSLGSPS GEVSHPRKTR TVVQPSVGAA AGPVVPPCPG RIRHFKV | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in PBS pH 7.4. |