missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Borrelia DbpB Protein
A cDNA sequence encoding the Borrelia DbpB was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11183-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Borrelia DbpB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Borrelia DbpB (1.11 mg/1ml) is supplied in 16mM HEPES buffer pH 8.0, 300mM NaCl and 20% glycerol. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |