missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DDAVP Protein
A cDNA sequence encoding the DDAVP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11607-5mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
DDAVP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Desmopressin peptide was lyophilized with no additives. |
5 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |