Learn More
enQuireBio™ Recombinant other Histrelin Protein
Recombinant Protein
Marca: enQuireBio™ QP12236-5mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Histrelin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Histrelin peptide was lyophilized with no additives. |
5 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 99.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |