missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Lysostaphin Protein
A cDNA sequence encoding the Lysostaphin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10778-5mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Lysostaphin Protein | |
Research Use Only | |
lss | |
Other | |
Untagged | |
The protein was lyophilized without any additives. |
5 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MKKTKNNYYT RPLAIGLSTF ALASIVYGGI QNETHASEKS NMDVSKKVAE VETSKAPVEN TAEVETSKAP VENTAEVETS KAPVENTAEV ETSKAPVENT AEVETSKAPV ENTAEVETSK APVENTAEVE TSKAPVENTA EVETSKAPVE NTAEVETSKA PVENTAEVET SKAPVENTAE VETSKAPVEN TAEVETSKAP VENTAEVETS KAPVENTAEV ETSKAPVENT AEVETSKALV QNRTALRAAT HEHSAQWLNN YKKGYGYGPY PLGINGGMHY GVDFFMNIGT PVKAISSGKI VEAGWSNYGG GNQIGLIEND GVHRQWYMHL SKYNVKVGDY VKAGQIIGWS GSTGYSTAPH LHFQRMVNSF SNSTAQDPMP FLKSAGYGKA GGTVTPTPNT GWKTNKYGTL YKSESASFTP NTDIITRTTG PFRSMPQSGV LKAGQTIHYD EVMKQDGHVW VGYTGNSGQR IYLPVRTWNK STNTLGVLWG TIK | |
97% as determined by RP-HPLC. |