missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other PMSG Protein
A cDNA sequence encoding the PMSG was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13081-100000IU
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PMSG Protein | |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
The PMSG was lyophilized with no additives. |
100000 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |