missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other SODA Protein
A cDNA sequence encoding the SODA was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10867-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
SODA Protein | |
Research Use Only | |
Specific activity is > 350 units/mg, in which one unit will inhibit the rate of reduction of cytochrome c by 50% in a coupled system, using xanthine and xanthine oxidase at pH 7.8 at 25C in a 1.5 ml reaction volume. | |
Other | |
His | |
SODA E. coli solution (1 mg/ml) containining 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSYTLPSLPY AYDALEPHFD KQTMEIHHTK HHQTYVNNAN AALESLPEFA NLPVEELITK LDQLPADKKT VLRNNAGGHA NHSLFWKGLK KGTTLQGDLK AAIERDFGSV DNFKAEFEKA AASRFGSGWA WLVLKGDKLA VVSTANQDSP LMGEAISGAS GFPIMGLDVW EHAYYLKFQN RRPDYIKEFW NVVNWDEAAA RFAAKK | |
Greater than 95% as determined by SDS-PAGE. |