missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa L-ornithine N(5)-monooxygenase Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) L-ornithine N(5)-monooxygenase, was expressed in the hosts and tags indicated. option, ab
Marca: enQuireBio™ QP7676-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_251076.1 | |
P. aeruginosa L-ornithine N(5)-monooxygenase Protein | |
Research Use Only | |
pvdA | |
Recombinant Protein | |
E. coli | |
MTQATATAVV HDLIGVGFGP SNIALAIALQ ERAQAQGALE VLFLDKQGDY RWHGNTLVSQ SELQISFLKD LVSLRNPTSP YSFVNYLHKH DRLVDFINLG TFYPCRMEFN DYLRWVASHF QEQSRYGEEV LRIEPMLSAG QVEALRVISR NADGEELVRT TRALVVSPGG TPRIPQVFRA LKGDGRVFHH SQYLEHMAKQ PCSSGKPMKI AIIGGGQSAA EAFIDLNDSY PSVQADMILR ASALKPADDS PFVNEVFAPK FTDLIYSREH AERERLLREY HNTNYSVVDT DLIERIYGVF YRQKVSGIPR HAFRCMTTVE RATATAQGIE LALRDAGSGE LSVETYDAVI LATGYERQLH RQLLEPLAEY LGDHEIGRDY RLQTDERCKV AIYAQGFSQA SHGLSDTLLS VLPVRAEEIS GSLYQHLKPG TAARALHEHA LAS | |
Greater than 90% as determined by SDS-PAGE. |
882167 | |
500 μg | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |