missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Erythropoietin / EPO Protein
A DNA sequence encoding the Sus scrofa (Pig) Erythropoietin / EPO, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5990-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_999299.1 | |
Erythropoietin / EPO Protein | |
Research Use Only | |
EPO | |
Recombinant Protein | |
Yeast | |
APPRLICDSR VLERYILEAK EGENATMGCA ESCSFSENIT VPDTKVNFYA WKRMEVQQQA MEVWQGLALL SEAILQGQAL LANSSQPSEA LQLHVDKAVS GLRSLTSLLR ALGAQKEAIP LPDASPSSAT PLRTFAVDTL CKLFRNYSNF LRGKLTLYTG EACRRRDR | |
Greater than 90% as determined by SDS-PAGE. |
397249 | |
50 μg | |
Not determined. | |
Full Length | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |