missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine LH Protein
A cDNA sequence encoding the LH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12568-750IU
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
397153 | |
750 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Porcine | |
MEMLQGLLLW LLLSVAGVWA SRGPLRPLCR PINATLAAEN EACPVCITFT TSICAGYCPS MVRVLPAALP PVPQPVCTYR ELSFASIRLP GCPPGVDPTV SFPVALSCHC GPCRLSSSDC GGPRAQPLAC DRPLLPGLLF L |
LH Protein | |
Research Use Only | |
LHB | |
Porcine | |
Untagged | |
The LH was lyophilized with no additives. |