missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL24 / Eotaxin-2 / MPIF-2 Protein
A DNA sequence encoding the Rat CCL24 / Eotaxin-2 / MPIF-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10299-ec-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001013063.1 | |
CCL24 / Eotaxin-2 / MPIF-2 Protein | |
Research Use Only | |
CCL24 | |
The biologically active determined by a chemotaxis bioassay using human peripheral blood eosinophils is in a concentration of 50-250 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
288593 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MPTGSVTIPS SCCVTFISKK IPVNRVISYQ LANGSICPKA GVIFITKKGH KICTDPKLPW VQKHIKNLDA KRNQPSEGAK ALGPKFVIQK LRGNSTKV | |
Greater than 96% as determined by SDS-PAGE and HPLC. |