missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CXCL12 / SDF-1 Isoform 1 Protein
A DNA sequence encoding the Rat CXCL12 / SDF-1 Isoform 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10287-ec-250ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_071513.1 | |
CXCL12 / SDF-1 Isoform 1 Protein | |
Research Use Only | |
CXCL12 | |
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 50-100 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
24772 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKSNNRQVC IDPKLKWIQE YLDKALNK | |
Greater than 97% as determined by SDS-PAGE and HPLC. |