missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CXCL2 / MIP-2 Protein
A DNA sequence encoding the Rat CXCL2 / MIP-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10281-ec-25ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_446099.1 | |
CXCL2 / MIP-2 Protein | |
Research Use Only | |
CXCL2 | |
The biological activity determined by a chemotaxis bioassay using rat neutrophils is in a concentration range of 10-100 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
114105 | |
25 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
VVVASELRCQ CLTTLPRVDF KNIQSLTVTP PGPHCAQTEV IATLKDGHEV CLNPEAPLVQ RIVQKILNKG KAN | |
Greater than 98% as determined by SDS-PAGE and HPLC. |