missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Disks large homolog 3 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Disks large homolog 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7691-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_113827.1 | |
Disks large homolog 3 Protein | |
Research Use Only | |
Dlg3 | |
Recombinant Protein | |
E. coli | |
MHKHQHCCKC PECYEVTRLA ALRRLEPPGY GDWQVPDPYG PSGGNGASSG YGGYSSQTLP SQAGATPTPR TKAKLIPTGR DVGPVPPKPV PGKNTPKLNG SGPSWWPECT CTNRDWYEQA SPAPLLVNPE ALEPSLSVNG SDGMFKYEEI VLERGNSGLG FSIAGGIDNP HVPDDPGIFI TKIIPGGAAA MDGRLGVNDC VLRVNEVDVS EVVHSRAVEA LKEAGPVVRL VVRRRQPPPE TIMEVNLLKG PKGLGFSIAG GIGNQHIPGD NSIYITKIIE GGAAQKDGRL QIGDRLLAVN NTNLQDVRHE EAVASLKNTS DMVYLKVAKP GSLHLNDMYA PPDYASTFTA LADNHISHNS SLGYLGAVES KVTYPAPPQV PPTRYSPIPR HMLAEEDFTR EPRKIILHKG STGLGFNIVG GEDGEGIFVS FILAGGPADL SGELRRGDRI LSVNGVNLRN ATHEQAAAAL KRAGQSVTIV AQYRPEEYSR FESKIHDLRE QMMNSSMSSG SGSLRTSEKR SLYVRALFDY DRTRDSCLPS QGLSFSYGDI LHVINASDDE WWQARLVTPH GESEQIGVIP SKKRVEKKER ARLKTVKFHA RTGMIESNRD FPGLSDDYYG AKNLKGVTSN TSDSESSSKG QEDAILSYEP VTRQEIHYAR PVIILGPMKD RVNDDLISEF PHKFGSCVPH TTRPRRDNEV DGQDYHFVVS REQMEKDIQD NKFIEAGQFN DNLYGTSIQS VRAVAERGKH CILDVSGNAI KRLQQAQLYP IAIFIKPKSI EALMEMNRRQ TYEQANKIFD KAMKLEQEFG EYFTAIVQGD SLEEIYNKIK QIIEDQSGHY IWVPSPEKL | |
Greater than 90% as determined by SDS-PAGE. |
58948 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |