missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat HB-EGF Protein
A cDNA sequence encoding the HB-EGF was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10663-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
HB-EGF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
DLEGTDLDLF KVAFSSKPQA LATPGKEKNG KKKRKGKGLG KKRDPCLKKY KDYCIHGECR YLKELRIPSC HCLPGYHGQR CHGLTL | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Rat | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution containing 1xPBS pH 7.4. |