missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Lumican / LUM Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Lumican / LUM, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6324-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_112312.1 | |
Lumican / LUM Protein | |
Research Use Only | |
Lum | |
Recombinant Protein | |
E. coli | |
QYYDYDAPLF MYGELSPNCA PECNCPHSYP TAMYCDDLKL KSVPMVPPGI KYLYLRNNQI DHIDEKAFEN VTDLQWLILD HNLLENSKIK GKVFSKLKQL KKLHINYNNL TESVGPLPKS LQDLQLANNK ISKLGSFDGL VNLTFIYLQH NQLKEEAVSA SLKGLKSLEY LDLSFNQMSK LPAGLPTSLL TLYLDNNKIT NIPDEYFNRF TGLQYLRLSH NELADSGVPG NSFNISSLLE LDLSYNKLKS IPTVNENLEN YYLEVNKLEK FDVKSFCKIL GPLSYSKIKH LRLDGNPLTQ SSLPPDMYEC LRVANEITVN | |
Greater than 90% as determined by SDS-PAGE. |
81682 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |