Learn More
enQuireBio™ Recombinant Rat M-CSF / CSF-1 Protein
Recombinant Protein
Marca: enQuireBio™ QP5518-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
M-CSF / CSF-1 Protein | |
Research Use Only | |
CSF1 | |
Recombinant Protein | |
E. coli | |
MEVSEHCSHM IGNGHLQILQ QLIDSQMETA CLIEYKFVDQ EQLDDPVCYL KKAFVLVQVI IEETMRFKDN TPNANATERL QELSMKLNSC FIKDYKEQNE ACVQTYKESP LRLLEKIKNF FNETKNFLEK DWNIFSKNCN DSLAKCSSRD VVTKP | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat M-CSF / CSF-1 induces NFS-60 cell proliferation (typical ED50 is ? 2 ng/ml). | |
Rat | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |