missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat MMP7 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) MMP7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6371-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_036996.1 | |
MMP7 Protein | |
Research Use Only | |
Mmp7 | |
Recombinant Protein | |
Yeast | |
FSLMPNSPKW HSRTVTYRIV SYTTDLPRFL VDQIVKRALR MWSMQIPLNF KRVSWGTADI IIGFARGDHG DNFPFDGPGN TLGHAFAPGP GLGGDAHFDK DEYWTDGEDS GVNFLFVATH ELGHSLGLGH SSVPSSVMYP TYQGDHSEDF SLTKDDIAGI QKLYGKRNKL | |
Greater than 90% as determined by SDS-PAGE. |
25335 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |