missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Plasma kallikrein Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Plasma kallikrein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6269-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Plasma kallikrein Protein | |
Research Use Only | |
Klkb1 | |
Recombinant Protein | |
E. coli | |
IVGGTNSSLG EWPWQVSLQV KLVSQNHMCG GSIIGRQWIL TAAHCFDGIP YPDVWRIYGG ILNLSEITNK TPFSSIKELI IHQKYKMSEG SYDIALIKLQ TPLNYTEFQK PICLPSKADT NTIYTNCWVT GWGYTKERGE TQNILQKATI PLVPNEECQK KYRDYVITKQ MICAGYKEGG IDACKGDSGG PLVCKHSGRW QLVGITSWGE GCARKEQPGV YTKVAEYIDW ILEKIQSSKE RALETSPA | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |