missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PLAUR / CD87 / uPAR Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PLAUR / CD87 / uPAR, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9334-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PLAUR / CD87 / uPAR Protein | |
Research Use Only | |
PLAUR | |
Recombinant Protein | |
Yeast | |
ASDESNCGCQ NGGVCVSYKY FSSIRRCSCP KKFKGEHCEI DTSKTCYHGN GQSYRGKANT DTKGRPCLAW NSPAVLQQTY NAHRSDALSL GLGKHNYCRN PDNQRRPWCY VQIGLKQFVQ ECMVQDCSLS KKPSSTVDQQ GFQCGQKALR PRFKIVGGEF TVVENQPWFA AIYLKNKGGS PPSFKCGGSL ISPCWVASAT HCFVNQPKKE EYVVYLGQSK RNSYNPGEMK FEVEQLILHE DFSDETLAFH NDIALLKIRT STGQCAQPSR TIQTI | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |