missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PLBD2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PLBD2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7660-ye-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_640348.2 | |
PLBD2 Protein | |
Research Use Only | |
Plbd2 | |
Recombinant Protein | |
Yeast | |
GALPTLGPGW RRQNPEPPAS RTRSLLLDAA SGQLRLEYGF HPDAVAWANL TNAIRETGWA YLDLGTNGSY NDSLQAYAAG VVEASVSEEL IYMHWMNTVV NYCGPFEYEV GYCEKLKSFL EANLEWMQRE MELSPDSPYW HQVRLTLLQL KGLEDSYEGR LTFPTGRFNI KPLGFLLLQI SGDLEDLEPA LNKTNTKPSV GSGSCSALIK LLPGSHDLLV AHNTWNSYQN MLRIIKKYRL QFREGPQEEY PLIAGNNLIF SSYPGTIFSG DDFYILGSGL VTLETTIGNK NPALWKYVQP QGCVLEWIRN IVANRLALDG ATWADVFRRF NSGTYNNQWM IVDYKAFIPN GPSPGSRVLT ILEQIPGMVV VADKTAELYK TTYWASYNIP YFESVFNASG LQALVAQYGD WFSYTRNPRA KIFQRDQSLV EDVDTMVRLM RYNDFLHDPL SLCEACSPKP NAENAISARS DLNPANGSYP FQALRQRAHG GIDVKVTSVA LAKYMSMLAA SGPTWDQLPP FQWSKSPFHN MLHMGQPDLW MFSPVKVPWD | |
Greater than 90% as determined by SDS-PAGE. |
246120 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |