missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PRL R Protein
A cDNA sequence encoding the PRL R was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10845-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PRL R Protein | |
Research Use Only | |
Activity is determined by the dose-dependant inhibition of Prolactin-stimuled proliferation of Nb2 cells and by high affinity binding of oPLR and other lactogenic hormones. | |
Rat | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MPSALAFVLL VLNISLLKGQ SPPGKPEIHK CRSPDKETFT CWWNPGTDGG LPTNYSLTYS KEGEKTTYEC PDYKTSGPNS CFFSKQYTSI WKIYIITVNA TNQMGSSSSD PLYVDVTYIV EPEPPRNLTL EVKQLKDKKT YLWVKWSPPT ITDVKTGWFT MEYEIRLKPE EAEEWEIHFT GHQTQFKVFD LYPGQKYLVQ TRCKPDHGYW SRWSQESSVE MPNDFTLKDT TVWIIVAILS AVICLIMVWA VALKGYSMMT CIFPPVPGPK IKGFDTHLLE KGKSEELLSA LGCQDFPPTS DCEDLLVEFL EVDDNEDERL MPSHSKEYPG QGVKPTHLDP DSDSGHGSYD SHSLLSEKCE EPQAYPPTLH IPEITEKPEN PEANIPPTVD PQSTNPNFHV DAPKSSTWPL LPGQHMPRSP YHSVADVCKL AGSPVNTLDS FLDKAEENVL KLSKALETGE EEVAEQKGAK SFPSDKQNTP WPLLQEKSPT VYVKPPDYVE IHKVNKDGVL SLFPKQRENN QTEKPGVPET SKEYAKVSGI TDNNILVLVP DSRAQNTALL EESAKKAPPS FEADQSEKDL ASFTATSSNR RLQLGRLDYL DPTCFMHSFH | |
Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |