missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat REG3A / HIP Protein
A DNA sequence encoding the Rattus norvegicus (Rat) REG3A / HIP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6594-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001139318.1 | |
REG3A / HIP Protein | |
Research Use Only | |
REG3A | |
Recombinant Protein | |
E. coli | |
EDSQKAVPST RTSCPMGSKA YRSYCYTLVT TLKSWFQADL ACQKRPSGHL VSILSGGEAS FVSSLVTGRV NNNQDIWIWL HDPTMGQQPN GGGWEWSNSD VLNYLNWDGD PSSTVNRGNC GSLTATSEFL KWGDHHCDVE LPFVCKFKQ | |
Greater than 90% as determined by SDS-PAGE. |
171162 | |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |