missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus 30 kDa neutral phosphatase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Oxygen-insensitive NADPH nitroreductase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7091-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
S. aureus 30 kDa neutral phosphatase Protein | |
Research Use Only | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
500 μg | |
Not determined. | |
Recombinant Protein | |
E. coli | |
KSSAEVQQTQ QASIPASQKA NLGNQNNIMX VAXYQ | |
Greater than 90% as determined by SDS-PAGE. |