missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Clumping factor A Protein
A DNA sequence encoding the Staphylococcus aureus (strain COL) Clumping factor A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7668-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
WP_001056223.1 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). Protein | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |
S. aureus Clumping factor A Protein | |
Research Use Only | |
clfA | |
Recombinant Protein | |
Yeast | |
GTDITNQLTN VTVGIDSGTT VYPHQAGYVK LNYGFSVPNS AVKGDTFKIT VPKELNLNGV TSTAKVPPIM AGDQVLANGV IDSDGNVIYT FTDYVNTKDD VKATLTMPAY IDPENVKKTG NVTLATGIGS TTANKTVLVD YEKYGKFYNL SIKGTIDQID KTNNTYRQTI YVNPSGDNVI APVLTGNLKP NTDSNALIDQ QNTSIKVYKV DNAADLSESY FVNPENFEDV TNSVNITFPN PNQYKVEFNT PDDQITTPYI VVVNGHIDPN SKGDLALRST LYGYNSNIIW RSMSWDNEVA FNNGSGSGDG IDKPVVPEQP DEPGEIEPIP E | |
Greater than 95% as determined by SDS-PAGE. |