missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Clumping factor B Protein
A DNA sequence encoding the Staphylococcus aureus (strain MRSA252) Clumping factor B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7759-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
WP_000745917.1 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus Clumping factor B Protein | |
Research Use Only | |
clfB | |
Recombinant Protein | |
E. coli | |
SEQSNDTTQS SKNNASADSE KNNTIETPQL NTTANDTSDI SANTNSANVD STAKTMSTQT SNTTTTEPAS TNETPQPTAI KDQATAAKMQ DQTVPQEANS QVDNKTTNDA NNIATNSELK NPQTLDLPQS SPQTISNAQG TSKPSVRTRA VRSLAVAEPV VNAADAKGTN VNDKVTASDF KLEKTAFDPN QSGNTFMAAN FKVTGQVKSG DYFTAKLPDS VTGNGDVDYS NSNNTMPIAD IKSTNGDVVA KATYDILTKT YTFVFTDYVN DKENINGQFS LPLFTDRAKA PKSGTYDANI NIADEMFDNK ITYNYSSPIA GIDKPNGANI SSQIIGVDTA SGQNTYKQTV FVNPKQRVLG NTWVYIKGYQ DKIEESSGKV SATDTKLRIF EVNDTSKLSD SYYADPNDSN LKEVTGEFKD KISYKYDNVA SINFGDINKT YVVLVEGHYD NTGKNLKTQV IQENIDPATG KDYSIFGWNN ENVVRYGGGS ADGDSAVN | |
Greater than 90% as determined by SDS-PAGE. |