missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Trehalose-phosphatase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Trehalose-phosphatase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8863-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_010359.1 | |
S. cerevisiae Trehalose-phosphatase Protein | |
Research Use Only | |
TPS2 | |
Recombinant Protein | |
E. coli | |
GEFHAKELKE KLLSFTDDFD LEVMDGKANI EVRPRFVNKG EIVKRLVWHQ HGKPQDMLKG ISEKLPKDEM PDFVLCLGDD FTDEDMFRQL NTIETCWKEK YPDQKNQWGN YGFYPVTVGS ASKKTVAKAH LTDPQQVLET LGLLVGDVSL FQSAGTVDLD SRGHVKNSES SLKSKLASKA YVMKRSASYT GAK | |
Greater than 90% as determined by SDS-PAGE. |
851646 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |