missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep IL6 / Interleukin-6 Protein
A DNA sequence encoding the Ovis aries (Sheep) IL6 / Interleukin-6 Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6225-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001009392.1 | |
IL6 / Interleukin-6 Protein | |
Research Use Only | |
IL6 | |
Recombinant Protein | |
Yeast | |
GPLGEDFKND TTPSRLLLTT PEKTEALIKH IVDKISAIRK EICEKNDECE NSKETLAENK LKLPKMEEKD GCFQSGFNQA ICLIKTTAGL LEYQIYLDFL QNEFEGNQET VMELQSSIRT LIQILKEKIA GLITTPATHT DMLEKMQSSN EWVKNAKVII ILRSLENFLQ FSLRAIRMK | |
Greater than 80% as determined by SDS-PAGE. |
443406 | |
50 μg | |
Not determined. | |
Full Length | |
Sheep | |
His | |
Tris-based buffer, 50% glycerol |