Learn More
Fisherbrand™ Rectangular Basket for Autoclaves
For use with HMT 260 FA/MA and HMT 230 FA/MA Autoclaves.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 2.00000kg
Descrizione
- Solid construction
- Wire mesh basket
- 370 x 210 x 105mm or 340 x 180 x 120mm
Specifica
Autoclavable | |
Basket, Rectangular | |
340 x 180 x 120 mm |
HMT 230 FA/MA Tabletop Autoclaves | |
Wire Mesh |