Learn More
Brady™ Reinforced Polypropylene Mandatory Signs
Marca: Brady™ 252114
Dettagli aggiuntivi : Peso : 0.13000kg
Specifica
Polypropylene | |
Reinforced Polypropylene | |
120°C | |
Round | |
Mandatory Sign |
1 Ea. | |
315 mm | |
0.369 mm | |
Protective Gloves | |
White on Blue |