Learn More
RELL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553051
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: NEANADVLKA MVADNSLYDP ESPVTPSTPG SPPVSPGPLS PGGTPGKHVC GHHLHTVGGV VERDVCHRCR HKRWHFIKPT NKSRESRPRR QGEVTVLSVG RFRVTKVEHK SNQKERRSLM SVSGAETVNG E Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
RELL1 Polyclonal antibody specifically detects RELL1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifica
RELL1 | |
Polyclonal | |
Unconjugated | |
RELL1 | |
AA536743; PSEC0162; receptor expressed in lymphoid tissues like 1; RELL1; RELT like 1; RELT-like 1; RELT-like protein 1; RGD1307468; tmp_locus_29 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
768211 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IUW5 | |
RELL1 | |
Recombinant protein corresponding to Human RELL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only