Learn More
Labconco™ Glass Manifold
Make exhaustion of fumes from Kjeldahl digestions more convenient by adding a Labconco Glass Manifold to your system.
Marca: Labconco™ 6001100
Dettagli aggiuntivi : Peso : 2.00000kg
Descrizione
- Six-port glass manifold exhausts fumes from Kjeldahl digestions
- Serrated connector for water aspirator
Specifica
Manifold | |
Glass |